siRNA Details
| Gene Symbol: Napsa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031670.2 | NP_113858.1 | 3 | 355 | ||
| GenBank mRNA | AJ251299.1 | - | 3 | 327 | ||
| BC078790.1 | - | 3 | 347 | 7109515 | ||
| FQ225885.1 | - | 3 | 333 | |||
| FQ230345.1 | - | 3 | 348 | |||

Napsa
Genomic Map