siRNA Details
| Gene Symbol: Ctla4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031674.1 | NP_113862.1 | 2 | 268 | ||
| XM_006245036.2 | XP_006245098.1 | 2 | 386 | |||
| GenBank mRNA | U37121.1 | - | 2 | 268 | ||
| U90271.1 | - | 2 | 268 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ctla4| Gene Symbol: Ctla4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031674.1 | NP_113862.1 | 2 | 268 | ||
| XM_006245036.2 | XP_006245098.1 | 2 | 386 | |||
| GenBank mRNA | U37121.1 | - | 2 | 268 | ||
| U90271.1 | - | 2 | 268 | |||