siRNA Details
| Gene Symbol: Fhl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031677.1 | NP_113865.1 | 3 | 427 | ||
| XM_006244849.2 | XP_006244911.1 | 4 | 2564 | |||
| GenBank mRNA | AB008571.1 | - | 3 | 427 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fhl2| Gene Symbol: Fhl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031677.1 | NP_113865.1 | 3 | 427 | ||
| XM_006244849.2 | XP_006244911.1 | 4 | 2564 | |||
| GenBank mRNA | AB008571.1 | - | 3 | 427 | ||