siRNA Details
| Gene Symbol: Syt4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031693.2 | NP_113881.1 | 2 | 444 | ||
| GenBank mRNA | BG666223.1 | - | 2 | 210 | ||
| CB786178.1 | - | 2 | 434 | |||
| DQ181551.1 | - | 2 | 208 | |||
| L38247.1 | - | 2 | 445 | |||
| U14398.1 | - | 2 | 473 | |||

Syt4
Genomic Map