siRNA Details
| Gene Symbol: Gpr88 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031696.1 | NP_113884.1 | 2 | 2002 | ||
| XM_006233220.2 | XP_006233282.1 | 2 | 1915 | |||
| XM_006233221.2 | XP_006233283.1 | 2 | 1860 | |||
| XM_006233222.2 | XP_006233284.1 | 2 | 1857 | |||
| XM_008761471.1 | XP_008759693.1 | 3 | 2474 | |||
| GenBank mRNA | AB042407.1 | - | 2 | 2002 | ||
More Information
Set Membership: |
|

Gpr88
Genomic Map