siRNA Details
| Gene Symbol: St3gal3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031697.1 | NP_113885.1 | 5 | 314 | ||
| XM_006238715.2 | XP_006238777.1 | 5 | 751 | |||
| XM_006238716.2 | XP_006238778.1 | 4 | 717 | |||
| XM_008764066.1 | XP_008762288.1 | 5 | 3241 | |||
| XM_008764067.1 | XP_008762289.1 | 5 | 334 | |||
| GenBank mRNA | M97754.1 | - | 5 | 314 | ||

St3gal3
Genomic Map