siRNA Details
| Gene Symbol: Dpys | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031705.1 | NP_113893.1 | 2 | 439 | ||
| GenBank mRNA | BC081768.1 | - | 2 | 394 | 7107549 | |
| D63704.1 | - | 2 | 439 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dpys| Gene Symbol: Dpys | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031705.1 | NP_113893.1 | 2 | 439 | ||
| GenBank mRNA | BC081768.1 | - | 2 | 394 | 7107549 | |
| D63704.1 | - | 2 | 439 | |||