siRNA Details
| Gene Symbol: Adrm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031708.1 | NP_113896.1 | 4 | 433 | ||
| XM_006235822.1 | XP_006235884.1 | 4 | 554 | |||
| GenBank mRNA | AB032742.1 | - | 4 | 433 | ||
| BC061773.1 | - | 4 | 449 | 5621873 | ||
| FQ218007.1 | - | 4 | 451 | |||

Adrm1
Genomic Map