siRNA Details
| Gene Symbol: Hrsp12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031714.1 | NP_113902.1 | 6 | 735 | ||
| GenBank mRNA | AF015949.1 | - | 6 | 735 | ||
| BC078779.1 | - | 6 | 725 | 7128728 | ||
| D49363.1 | - | 6 | 704 | |||
| FQ209529.1 | - | 6 | 747 | |||
| FQ209945.1 | - | 5 | 584 | |||
| FQ210021.1 | - | 6 | 739 | |||
| FQ210226.1 | - | 1 | 325 | |||
| FQ218425.1 | - | 6 | 741 | |||
| FQ218811.1 | - | 6 | 742 | |||
| FQ219440.1 | - | 6 | 741 | |||
| FQ219496.1 | - | 6 | 747 | |||
| X70825.1 | - | 6 | 697 | |||

Hrsp12
Genomic Map