siRNA Details
| Gene Symbol: Scamp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031725.1 | NP_113913.1 | 4 | 230 | ||
| XM_006240999.1 | XP_006241061.1 | 4 | 249 | |||
| GenBank mRNA | AF285631.1 | - | 4 | 230 | ||
| BC090342.1 | - | 4 | 224 | 7304207 | ||

Scamp4
Genomic Map