siRNA Details
| Gene Symbol: Limk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031727.2 | NP_113915.2 | 16 | 2722 | ||
| XM_006249185.2 | XP_006249247.1 | 15 | 5814 | |||
| GenBank mRNA | D31873.1 | - | 16 | 2747 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Limk1| Gene Symbol: Limk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031727.2 | NP_113915.2 | 16 | 2722 | ||
| XM_006249185.2 | XP_006249247.1 | 15 | 5814 | |||
| GenBank mRNA | D31873.1 | - | 16 | 2747 | ||