siRNA Details
| Gene Symbol: Kcnd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031730.2 | NP_113918.2 | 6 | 2347 | ||
| XM_008762830.1 | XP_008761052.1 | 6 | 871 | |||
| GenBank mRNA | CB790894.1 | - | 2 | 318 | ||
| M59980.1 | - | 6 | 2024 | |||
| S64320.1 | - | 6 | 2345 | |||

Kcnd2
Genomic Map