siRNA Details
| Gene Symbol: Stk3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031735.1 | NP_113923.1 | 6 | 667 | ||
| XM_008765458.1 | XP_008763680.1 | 6 | 845 | |||
| XM_008765459.1 | XP_008763681.1 | 6 | 845 | |||
| XM_008765460.1 | XP_008763682.1 | 6 | 534 | |||
| GenBank mRNA | AJ001529.1 | - | 6 | 667 | ||
| BC161844.1 | - | 6 | 750 | 9027884 | ||

Stk3
Genomic Map