siRNA Details
| Gene Symbol: Slc2a5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031741.1 | NP_113929.1 | 12 | 2129 | ||
| GenBank mRNA | BC161843.1 | - | 12 | 2116 | 7127817 | |
| D13871.1 | - | 12 | 2129 | |||
| L05195.1 | - | 12 | 2101 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc2a5| Gene Symbol: Slc2a5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031741.1 | NP_113929.1 | 12 | 2129 | ||
| GenBank mRNA | BC161843.1 | - | 12 | 2116 | 7127817 | |
| D13871.1 | - | 12 | 2129 | |||
| L05195.1 | - | 12 | 2101 | |||