siRNA Details
| Gene Symbol: CTRC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007272.2 | NP_009203.2 | 6 | 615 | ||
| GenBank mRNA | AK289447.1 | - | 6 | 614 | ||
| BC015118.1 | - | 6 | 621 | 3950170 | ||
| S82198.1 | - | 6 | 592 | |||
| Y13697.1 | - | 5 | 544 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CTRC| Gene Symbol: CTRC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007272.2 | NP_009203.2 | 6 | 615 | ||
| GenBank mRNA | AK289447.1 | - | 6 | 614 | ||
| BC015118.1 | - | 6 | 621 | 3950170 | ||
| S82198.1 | - | 6 | 592 | |||
| Y13697.1 | - | 5 | 544 | |||