siRNA Details
Gene Symbol: Ggcx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031756.1 | NP_113944.1 | 6, 7 | 809 | ||
GenBank mRNA | AF065387.1 | - | 6, 7 | 809 | ||
BC168647.1 | - | 6, 7 | 780 | 9102306 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ggcx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031756.1 | NP_113944.1 | 6, 7 | 809 | ||
GenBank mRNA | AF065387.1 | - | 6, 7 | 809 | ||
BC168647.1 | - | 6, 7 | 780 | 9102306 |