siRNA Details
| Gene Symbol: Rabac1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031774.1 | NP_113962.1 | 4, 5 | 570 | ||
| GenBank mRNA | AF025506.1 | - | 4, 5 | 570 | ||
| BC086387.1 | - | 4, 5 | 537 | 7302383 | ||
| FQ210487.1 | - | 4, 5 | 563 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rabac1| Gene Symbol: Rabac1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031774.1 | NP_113962.1 | 4, 5 | 570 | ||
| GenBank mRNA | AF025506.1 | - | 4, 5 | 570 | ||
| BC086387.1 | - | 4, 5 | 537 | 7302383 | ||
| FQ210487.1 | - | 4, 5 | 563 | |||