siRNA Details
Gene Symbol: Wfs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031823.1 | NP_114011.1 | 3, 4 | 488 | ||
GenBank mRNA | AF136378.1 | - | 3, 4 | 488 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Wfs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031823.1 | NP_114011.1 | 3, 4 | 488 | ||
GenBank mRNA | AF136378.1 | - | 3, 4 | 488 |