siRNA Details
| Gene Symbol: Flot2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001270800.1 | NP_001257729.1 | 2 | 186 | ||
| NM_031830.2 | NP_114018.1 | 2 | 186 | |||
| XM_006246952.1 | XP_006247014.1 | 2 | 723 | |||
| GenBank mRNA | AF023302.1 | - | 2 | 165 | ||
| AF023303.1 | - | 2 | 165 | |||
| BC083550.1 | - | 2 | 153 | 7133999 | ||
| CB581049.1 | - | 2 | 186 | |||

Flot2
Genomic Map