siRNA Details
Gene Symbol: Clta | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031974.1 | NP_114180.1 | 7 | 931 | ||
XM_006238104.1 | XP_006238166.1 | 6 | 1668 | |||
XM_006238105.1 | XP_006238167.1 | 6 | 1650 | |||
XM_006238106.1 | XP_006238168.1 | 5 | 1614 | |||
GenBank mRNA | BC087577.1 | - | 5 | 874 | 7307624 | |
FQ213034.1 | - | 7 | 939 | |||
FQ213766.1 | - | 6 | 896 | |||
FQ220908.1 | - | 5 | 849 | |||
FQ230032.1 | - | 5 | 842 | |||
M15882.1 | - | 7 | 931 | |||
M19260.1 | - | 6 | 895 | |||
M19261.1 | - | 5 | 841 |
