siRNA Details
| Gene Symbol: Crot | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031987.1 | NP_114193.1 | 4 | 409 | ||
| XM_006236004.2 | XP_006236066.1 | 4 | 569 | |||
| XM_006236005.2 | XP_006236067.1 | 5 | 1899 | |||
| GenBank mRNA | BC074004.1 | - | 4 | 429 | 7098081 | |
| J02844.1 | - | 850 | ||||
| U26033.1 | - | 4 | 409 | |||

Crot
Genomic Map