siRNA Details
| Gene Symbol: Cntnap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032061.2 | NP_114450.1 | 6 | 949 | ||
| XM_006247431.2 | XP_006247493.1 | 6 | 971 | |||
| XM_008768122.1 | XP_008766344.1 | 6 | 961 | |||
| XM_008768123.1 | XP_008766345.1 | 6 | 764 | |||
| XM_008768124.1 | XP_008766346.1 | 5 | 728 | |||
| GenBank mRNA | AF000114.1 | - | 6 | 937 | ||
| U87224.1 | - | 6 | 949 | |||

Cntnap1
Genomic Map