siRNA Details
| Gene Symbol: Cntnap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032061.2 | NP_114450.1 | 12 | 1992 | ||
| XM_006247431.2 | XP_006247493.1 | 12 | 2014 | |||
| XM_008768122.1 | XP_008766344.1 | 12 | 2004 | |||
| XM_008768123.1 | XP_008766345.1 | 12 | 1807 | |||
| XM_008768124.1 | XP_008766346.1 | 11 | 1771 | |||
| GenBank mRNA | AF000114.1 | - | 12 | 1980 | ||
| U87224.1 | - | 12 | 1992 | |||

Cntnap1
Genomic Map