siRNA Details
| Gene Symbol: Dll1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032063.2 | NP_114452.1 | 5 | 990 | ||
| GenBank mRNA | U78889.2 | - | 5 | 990 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dll1| Gene Symbol: Dll1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032063.2 | NP_114452.1 | 5 | 990 | ||
| GenBank mRNA | U78889.2 | - | 5 | 990 | ||