siRNA Details
| Gene Symbol: Ptger4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032076.3 | NP_114465.3 | 1 | 113 | ||
| XM_006231996.2 | XP_006232058.1 | 2 | 2682 | |||
| GenBank mRNA | D28860.1 | - | 2 | 293 | ||
| U94709.1 | - | 1 | 113 | |||
More Information
Other Species Targeted
Set Membership: |
|

Ptger4
Genomic Map