siRNA Details
| Gene Symbol: Dnaja2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032079.2 | NP_114468.2 | 3 | 214 | ||
| GenBank mRNA | BC087010.1 | - | 3 | 214 | 7191358 | |
| U95727.1 | - | 3 | 248 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dnaja2| Gene Symbol: Dnaja2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032079.2 | NP_114468.2 | 3 | 214 | ||
| GenBank mRNA | BC087010.1 | - | 3 | 214 | 7191358 | |
| U95727.1 | - | 3 | 248 | |||