siRNA Details
| Gene Symbol: Hao2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032082.2 | NP_114471.1 | 5 | 526 | ||
| XM_006233022.2 | XP_006233084.1 | 6 | 682 | |||
| XM_006233024.2 | XP_006233086.1 | 6 | 682 | |||
| XM_006233025.2 | XP_006233087.1 | 5 | 541 | |||
| GenBank mRNA | BC078781.1 | - | 4 | 442 | 7132928 | |
| X67156.1 | - | 5 | 527 | |||

Hao2
Genomic Map