siRNA Details
| Gene Symbol: Cdo1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_052809.1 | NP_434696.1 | 1 | 312 | ||
| XM_006254698.2 | XP_006254760.1 | 1 | 537 | |||
| GenBank mRNA | BC070509.1 | - | 1 | 303 | 7100321 | |
| M35266.1 | - | 1 | 312 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cdo1| Gene Symbol: Cdo1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_052809.1 | NP_434696.1 | 1 | 312 | ||
| XM_006254698.2 | XP_006254760.1 | 1 | 537 | |||
| GenBank mRNA | BC070509.1 | - | 1 | 303 | 7100321 | |
| M35266.1 | - | 1 | 312 | |||