siRNA Details
| Gene Symbol: Ccnh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_052981.2 | NP_443213.2 | 4 | 559 | ||
| GenBank mRNA | AF154914.1 | - | 3 | 256 | ||
| AJ276495.1 | - | 4 | 559 | |||
| BC059109.1 | - | 3 | 362 | 6886953 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ccnh| Gene Symbol: Ccnh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_052981.2 | NP_443213.2 | 4 | 559 | ||
| GenBank mRNA | AF154914.1 | - | 3 | 256 | ||
| AJ276495.1 | - | 4 | 559 | |||
| BC059109.1 | - | 3 | 362 | 6886953 | ||