siRNA Details
| Gene Symbol: Clcnka | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053327.1 | NP_445779.1 | 20 | 2362 | ||
| XM_008764291.1 | XP_008762513.1 | 20 | 2478 | |||
| XM_008764292.1 | XP_008762514.1 | 20 | 2448 | |||
| GenBank mRNA | BC081761.1 | - | 20 | 2428 | 7108273 | |
| D13927.1 | - | 20 | 2362 | |||

Clcnka
Genomic Map