siRNA Details
| Gene Symbol: Bhlhe40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053328.1 | NP_445780.1 | 4 | 643 | ||
| GenBank mRNA | AF009330.1 | - | 4 | 643 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Bhlhe40| Gene Symbol: Bhlhe40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053328.1 | NP_445780.1 | 4 | 643 | ||
| GenBank mRNA | AF009330.1 | - | 4 | 643 | ||