siRNA Details
| Gene Symbol: Ogfr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053340.2 | NP_445792.2 | 1, 2 | 171 | ||
| GenBank mRNA | BC101898.1 | - | 1, 2 | 152 | 7932932 | |
| EV778659.1 | - | 1, 2 | 171 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ogfr| Gene Symbol: Ogfr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053340.2 | NP_445792.2 | 1, 2 | 171 | ||
| GenBank mRNA | BC101898.1 | - | 1, 2 | 152 | 7932932 | |
| EV778659.1 | - | 1, 2 | 171 | |||