siRNA Details
| Gene Symbol: Cxxc4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008761530.1 | XP_008759752.1 | 3 | 1537 | ||
| GenBank mRNA | AF272158.1 | - | 2 | 600 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cxxc4| Gene Symbol: Cxxc4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008761530.1 | XP_008759752.1 | 3 | 1537 | ||
| GenBank mRNA | AF272158.1 | - | 2 | 600 | ||