siRNA Details
| Gene Symbol: Dclk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053343.3 | NP_445795.1 | 3 | 321 | ||
| XM_008761004.1 | XP_008759226.1 | 3 | 2226 | |||
| XM_008761005.1 | XP_008759227.1 | 3 | 1249 | |||
| XM_008761006.1 | XP_008759228.1 | 3 | 2229 | |||
| XM_008761007.1 | XP_008759229.1 | 3 | 333 | |||
| GenBank mRNA | CF978300.1 | - | 563 | |||
| FQ211799.1 | - | 3 | 311 | |||
| U78857.1 | - | 3 | 311 | |||

Dclk1
Genomic Map