siRNA Details
| Gene Symbol: COG2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145036.1 | NP_001138508.1 | 13 | 1575 | ||
| NM_007357.2 | NP_031383.1 | 13 | 1575 | |||
| GenBank mRNA | AK295251.1 | - | 13 | 1582 | ||
| AK302288.1 | - | 5 | 548 | |||
| AK302791.1 | - | 14 | 1759 | |||
| AK312678.1 | - | 13 | 1491 | |||
| BC014960.1 | - | 13 | 1538 | 4876271 | ||
| BC051906.1 | - | 13 | 1464 | 4541036 | ||
| Z34975.1 | - | 13 | 1519 | |||

COG2
Genomic Map