siRNA Details
| Gene Symbol: Dclk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053343.3 | NP_445795.1 | 13 | 1652 | ||
| XM_008761004.1 | XP_008759226.1 | 13 | 3554 | |||
| XM_008761006.1 | XP_008759228.1 | 14 | 3631 | |||
| XM_008761007.1 | XP_008759229.1 | 13 | 1664 | |||
| GenBank mRNA | FQ211799.1 | - | 14 | 1713 | ||
| U78857.1 | - | 13 | 1642 | |||

Dclk1
Genomic Map