siRNA Details
| Gene Symbol: Tcf4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053369.1 | NP_445821.1 | 9, 10 | 851 | ||
| GenBank mRNA | AF149284.1 | - | 9, 10 | 851 | ||
| FQ221525.1 | - | 605 | ||||
| FQ233045.1 | - | 5, 6 | 540 | |||
| U09228.1 | - | 3, 4 | 258 | |||

Tcf4
Genomic Map