siRNA Details
| Gene Symbol: Tcf4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053369.1 | NP_445821.1 | 17 | 1894 | ||
| GenBank mRNA | AF149284.1 | - | 17 | 1894 | ||
| FQ221525.1 | - | 1657 | ||||
| FQ226283.1 | - | 8 | 1033 | |||
| FQ227003.1 | - | 5 | 651 | |||
| FQ232404.1 | - | 5 | 650 | |||
| FQ233045.1 | - | 13 | 1526 | |||
| U09228.1 | - | 11 | 1313 | |||

Tcf4
Genomic Map