siRNA Details
| Gene Symbol: Timm10b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053371.2 | NP_445823.1 | 434 | |||
| GenBank mRNA | AF061242.1 | - | 4 | 571 | ||
| AF150106.1 | - | 435 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Timm10b| Gene Symbol: Timm10b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053371.2 | NP_445823.1 | 434 | |||
| GenBank mRNA | AF061242.1 | - | 4 | 571 | ||
| AF150106.1 | - | 435 | ||||