siRNA Details
| Gene Symbol: Atp1b4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053381.1 | NP_445833.1 | 4, 5 | 646 | ||
| XM_006257493.2 | XP_006257555.1 | 4, 5 | 937 | |||
| XM_006257494.2 | XP_006257556.1 | 4, 5 | 2371 | |||
| XM_006257495.2 | XP_006257557.1 | 4, 5 | 2356 | |||
| GenBank mRNA | AF158385.2 | - | 4, 5 | 646 | ||
| AF158386.2 | - | 4, 5 | 634 | |||

Atp1b4
Genomic Map