siRNA Details
| Gene Symbol: Dctn4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053404.2 | NP_445856.1 | 4, 5 | 435 | ||
| XM_006254783.2 | XP_006254845.1 | 4, 5 | 498 | |||
| GenBank mRNA | AF190798.1 | - | 4, 5 | 496 | ||
| AF192493.1 | - | 4, 5 | 435 | |||
| AF192494.1 | - | 4, 5 | 435 | |||
| BC100143.1 | - | 4, 5 | 440 | 7133708 | ||

Dctn4
Genomic Map