siRNA Details
| Gene Symbol: Maged1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053409.2 | NP_445861.1 | 3 | 382 | ||
| GenBank mRNA | AF217964.1 | - | 3 | 518 | ||
| BC081756.1 | - | 3 | 382 | 7134252 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Maged1| Gene Symbol: Maged1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053409.2 | NP_445861.1 | 3 | 382 | ||
| GenBank mRNA | AF217964.1 | - | 3 | 518 | ||
| BC081756.1 | - | 3 | 382 | 7134252 | ||