siRNA Details
| Gene Symbol: Snx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053411.1 | NP_445863.1 | 6, 7 | 729 | ||
| XM_006243317.2 | XP_006243379.1 | 6, 7 | 612 | |||
| XM_008766349.1 | XP_008764571.1 | 6, 7 | 784 | |||
| GenBank mRNA | AF218916.1 | - | 6, 7 | 729 | ||
| BC092201.1 | - | 6, 7 | 732 | 7309531 | ||

Snx1
Genomic Map