siRNA Details
| Gene Symbol: Ilf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053412.1 | NP_445864.1 | 4 | 416 | ||
| XM_008765990.1 | XP_008764212.1 | 4 | 551 | |||
| XM_008765991.1 | XP_008764213.1 | 4 | 552 | |||
| XM_008765992.1 | XP_008764214.1 | 4 | 536 | |||
| XM_008765993.1 | XP_008764215.1 | 3 | 512 | |||
| XM_008765994.1 | XP_008764216.1 | 3 | 497 | |||
| XM_008765995.1 | XP_008764217.1 | 4 | 551 | |||
| XM_008765996.1 | XP_008764218.1 | 4 | 536 | |||
| GenBank mRNA | AF220102.1 | - | 4 | 416 | ||

Ilf3
Genomic Map