siRNA Details
| Gene Symbol: Slc4a4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053424.1 | NP_445876.1 | 3 | 237 | ||
| XM_006250788.2 | XP_006250850.1 | 3 | 697 | |||
| XM_006250789.2 | XP_006250851.1 | 3 | 697 | |||
| XM_006250790.2 | XP_006250852.1 | 4 | 415 | |||
| XM_006250791.2 | XP_006250853.1 | 3 | 249 | |||
| GenBank mRNA | AF124441.1 | - | 3 | 237 | ||
| AF210250.1 | - | 3 | 244 | |||
| AF254802.1 | - | 3 | 237 | |||

Slc4a4
Genomic Map