siRNA Details
| Gene Symbol: Qsox1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109898.1 | NP_001103368.1 | 12 | 3247 | ||
| NM_053431.3 | NP_445883.1 | 13 | 2396 | |||
| GenBank mRNA | AF217799.1 | - | 13 | 2400 | ||
| AF285078.1 | - | 13 | 2417 | |||
| AY623665.1 | - | 12 | 3301 | |||

Qsox1
Genomic Map