siRNA Details
| Gene Symbol: Hamp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053469.1 | NP_445921.1 | 2 | 163 | ||
| GenBank mRNA | AF344185.1 | - | 2 | 163 | ||
| FQ222617.1 | - | 2 | 182 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hamp| Gene Symbol: Hamp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053469.1 | NP_445921.1 | 2 | 163 | ||
| GenBank mRNA | AF344185.1 | - | 2 | 163 | ||
| FQ222617.1 | - | 2 | 182 | |||