siRNA Details
| Gene Symbol: Cnga3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053495.1 | NP_445947.1 | 5 | 1436 | ||
| XM_008767042.1 | XP_008765264.1 | 7 | 1960 | |||
| XM_008767043.1 | XP_008765265.1 | 6 | 1846 | |||
| XM_008767044.1 | XP_008765266.1 | 6 | 1843 | |||
| GenBank mRNA | AB002801.1 | - | 5 | 1436 | ||
| AF031943.1 | - | 1 | 637 | |||
| AJ272428.1 | - | 7 | 1723 | |||
| AJ272429.1 | - | 6 | 1609 | |||
| U76221.1 | - | 1 | 64 | |||

Cnga3
Genomic Map