siRNA Details
| Gene Symbol: Prdx4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053512.2 | NP_445964.1 | 1, 2 | 266 | ||
| GenBank mRNA | AF106945.1 | - | 1, 2 | 267 | ||
| BC059122.1 | - | 1, 2 | 350 | 6918063 | ||
| FQ219258.1 | - | 1, 2 | 314 | |||
| FQ220216.1 | - | 1, 2 | 320 | |||
| FQ220257.1 | - | 1, 2 | 314 | |||
| FQ220425.1 | - | 1, 2 | 315 | |||
| FQ220916.1 | - | 1, 2 | 315 | |||
| FQ223235.1 | - | 1, 2 | 310 | |||
| FQ228535.1 | - | 1, 2 | 315 | |||

Prdx4
Genomic Map