siRNA Details
| Gene Symbol: Slc5a7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053521.1 | NP_445973.1 | 2 | 326 | ||
| XM_006244251.2 | XP_006244313.1 | 2 | 429 | |||
| XM_008766774.1 | XP_008764996.1 | 2 | 477 | |||
| XM_008766775.1 | XP_008764997.1 | 2 | 426 | |||
| GenBank mRNA | AB030947.1 | - | 2 | 326 | ||

Slc5a7
Genomic Map